2-Bromo-3,6-difluorobenzaldehyde 96%

Warning

H52320-03 H52320-MD
AAH52320-MDEA 85.52 USD
AAH52320-MD AAH52320-03
2-Bromo-3,6-difluorobenzaldehyde 96%
2-Bromo-3,6-difluorobenzaldehyde
Formula: C₇H₃BrF₂O
MW: 221.00 g/mol
Melting Pt: 49…51 °C
Storage Temperature: Ambient
MDL Number: MFCD12922618
CAS Number: 934987-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR