4'-Methoxybiphenyl-4-carboxylic acid 95%

Warning

Synonyms: 4'-Methoxy[1,1'-biphenyl]-4-carboxylic acid4'-Methoxy-[1,1'-biphenyl]-4-carboxylic acid

H52274-06 H52274-03
AAH52274-06EA 149.85 USD
AAH52274-06 AAH52274-03
4'-Methoxybiphenyl-4-carboxylic acid 95%
4'-Methoxybiphenyl-4-carboxylic acid
Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00185792
CAS Number: 725-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR