4-(2-(Dimethylamino)ethylcarbamoyl)phenylboronic acid pinacol ester 95%

Danger

Synonyms: N-(2-Dimethylaminoethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide4-(N-[2-(Dimethylamino)ethyl]carbamoyl)benzeneboronic acid pinacol ester

H52270-06 H52270-03
AAH52270-06EA 231.99 USD
AAH52270-06 AAH52270-03
4-(2-(Dimethylamino)ethylcarbamoyl)phenylboronic acid pinacol ester 95%
4-(2-(Dimethylamino)ethylcarbamoyl)phenylboronic acid pinacol ester
Formula: C₁₇H₂₇BN₂O₃
MW: 318.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864307
CAS Number: 832114-11-1
UN: 3259
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR