2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-1H-isoindole-1,3(2H)-dione 95%

Synonyms: 4-(Phthalimidomethyl)benzeneboronic acid, pinacol ester2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)isoindoline-1,3-dione

H52267-03 H52267-06
AAH52267-03EA 37.88 USD
AAH52267-03 AAH52267-06
2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-1H-isoindole-1,3(2H)-dione 95%
2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-1H-isoindole-1,3(2H)-dione
Formula: C₂₁H₂₂BNO₄
MW: 363.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179489
CAS Number: 138500-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR