4-Ethylbenzamidoxime 97%

Warning

H52259-06 H52259-03
AAH52259-06EA 584.03 USD
AAH52259-06 AAH52259-03
4-Ethylbenzamidoxime 97%
4-Ethylbenzamidoxime
Formula: C₉H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD07366399
CAS Number: 65264-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR