2-Bromo-4-phenylthiazole 97%

Warning

H52245-06 H52245-03
AAH52245-06EA 450.11 USD
AAH52245-06 AAH52245-03
2-Bromo-4-phenylthiazole 97%
2-Bromo-4-phenylthiazole
Formula: C₉H₆BrNS
Storage Temperature: Ambient
MDL Number: MFCD09746507
CAS Number: 57516-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR