2-Amino-4,5-dimethoxythiobenzamide 97%

Warning

H52243-03 H52243-06 H52243-MD
AAH52243-06EA 790.32 USD
AAH52243-06 AAH52243-03 AAH52243-MD
2-Amino-4,5-dimethoxythiobenzamide 97%
2-Amino-4,5-dimethoxythiobenzamide
Formula: C₉H₁₂N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD16658867
CAS Number: 1379207-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR