2-Amino-5-nitrothiobenzamide 97%

Warning

H52237-06 H52237-03
AAH52237-06EA 193.52 USD
AAH52237-06 AAH52237-03
2-Amino-5-nitrothiobenzamide 97%
2-Amino-5-nitrothiobenzamide
Formula: C₇H₇N₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD02258912
CAS Number: 25026-97-5
EINECS: 246-572-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR