3-Nitrobenzamidoxime 97%

Supplier: Thermo Scientific Chemicals
Warning

H52212-06 H52212-03
AAH52212-03EA 51.22 USD
AAH52212-03 AAH52212-06
3-Nitrobenzamidoxime 97%
3-Nitrobenzamidoxime
Formula: C₇H₇N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD00053609
CAS Number: 5023-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR