3,4-Dimethoxybenzamidoxime 97%

Warning

H52203-06 H52203-03
AAH52203-06EA 418.86 USD
AAH52203-06 AAH52203-03
3,4-Dimethoxybenzamidoxime 97%
3,4-Dimethoxybenzamidoxime
Formula: C₉H₁₂N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD06147747
CAS Number: 40312-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR