(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid 95%

Supplier: Thermo Scientific Chemicals

Synonyms: (1R,3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid(1S,3R)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid(1S,3R)-3-(Boc-amino)cyclopentanecarboxylic Acid

H52194-03 H52194-MD
AAH52194-MDEA 151.11 USD
AAH52194-MD AAH52194-03
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid 95%
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid
Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD01320857
CAS Number: 261165-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR