2-Benzyl-N-boc-D-proline 95%

Warning

H52186-MD H52186-03
AAH52186-MDEA 150.12 USD
AAH52186-MD AAH52186-03
2-Benzyl-N-boc-D-proline 95%
2-Benzyl-N-boc-D-proline
Formula: C₁₇H₂₃NO₄
MW: 305.37 g/mol
Melting Pt: 146 °C
Storage Temperature: Ambient
MDL Number: MFCD06659153
CAS Number: 706806-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR