L-Allylglycine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-(-)-alpha-Allylglycine

H52097-03 H52097-MD
AAH52097-03EA 276.99 USD
AAH52097-03 AAH52097-MD
L-Allylglycine 95%
2-Allyl-L-glycine
Formula: C₅H₉NO₂
MW: 115.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002627
CAS Number: 16338-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR