2-Benzyl-L-proline hydrochloride 95%
Supplier: Thermo Scientific Chemicals
H52080-MD
H52080-03
AAH52080-MDEA
244.99
USD
AAH52080-MD
AAH52080-03
2-Benzyl-L-proline hydrochloride 95%
2-Benzyl-L-proline hydrochloride
Formula:
C₁₂H₁₅NO₂·HCl MW: 241.72 g/mol Melting Pt: 270…272 °C Storage Temperature: Ambient |
MDL Number:
MFCD16660381 CAS Number: 86116-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...