Boc-3,4-dichloro-D-phenylalanine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Boc-D-Phe(3,4-Cl2)-OH
, Boc-3,4-dichloro-D-Phe-OH
H52070-MD
H52070-03
H52070-06
AAH52070-MDEA
47.86
USD
AAH52070-MD
AAH52070-06
AAH52070-03
Boc-3,4-dichloro-D-phenylalanine 95%
Boc-3,4-dichloro-D-phenylalanine
Formula:
C₁₄H₁₇Cl₂NO₄ MW: 334.193 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00151887 CAS Number: 114873-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...