Boc-L-β-homoalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-β-HomoAla-OH

H52066-03 H52066-MD
AAH52066-03EA 280.31 USD
AAH52066-03 AAH52066-MD
Boc-L-β-homoalanine 95%
Boc-L-beta-homoalanine
Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00270345
CAS Number: 158851-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR