Boc-(R)-3-amino-5-phenylpentanoic acid 95%

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-5-phenylpentanoic acid

H52055-MD H52055-03
AAH52055-MDEA 103.67 USD
AAH52055-MD AAH52055-03
Boc-(R)-3-amino-5-phenylpentanoic acid 95%
Boc-(R)-3-amino-5-phenylpentanoic acid
Formula: C₁₆H₂₃NO₄
MW: 293.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01076262
CAS Number: 218608-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR