Fmoc-L-β-homophenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-B-HoPhe-OHFmoc-β-HomoPhe-OH

H52052-03 H52052-MD
AAH52052-MDEA 110.72 USD
AAH52052-MD AAH52052-03
Fmoc-L-β-homophenylalanine 95%
Fmoc-L-beta-homophenylalanine
Formula: C₂₅H₂₃NO₄
MW: 401.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01863055
CAS Number: 193954-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR