(R)-3-(Boc-amino)-3-(2-bromophenyl)propionic acid 98%

Synonyms: (R)-3-(2-Bromophenyl)-3-((tert-butoxycarbonyl)amino)propanoic acid

H52028-03 H52028-MD H52028-06
AAH52028-03EA 233.1 USD
AAH52028-03 AAH52028-MD AAH52028-06
(R)-3-(Boc-amino)-3-(2-bromophenyl)propionic acid 98%
(R)-3-(Boc-amino)-3-(2-bromophenyl)propionic acid
Formula: C₁₄H₁₈BrNO₄
MW: 344.20 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD03427961
CAS Number: 500789-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR