Boc-D-3-Pyridylalanine 95%

Synonyms: Boc-β-(3-pyridyl)-D-Ala-OHBoc-D-3-Pal-OH

H51985-03 H51985-06 H51985-MD
AAH51985-03EA 142.08 USD
AAH51985-03 AAH51985-06 AAH51985-MD
Boc-D-3-Pyridylalanine 95%
Boc-D-3-Pyridylalanine
Formula: C₁₃H₁₈N₂O₄
MW: 266.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00076901
CAS Number: 98266-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR