(S)-N-Boc-4-bromophenylalanine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Boc-p-bromo-Phe-OH
, Boc-Phe(4-Br)-OH
H51969-06
H51969-03
AAH51969-03EA
70.29
USD
AAH51969-03
AAH51969-06
(S)-N-Boc-4-bromophenylalanine 98%
(S)-N-Boc-4-bromophenylalanine
Formula:
C₁₄H₁₈BrNO₄ MW: 344.2 g/mol Melting Pt: 118…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00237571 CAS Number: 62129-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...