3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzeneboronic acid, pinacol ester

H51939-03 H51939-06
AAH51939-03EA 39.39 USD
AAH51939-03 AAH51939-06
3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester 95%
3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Formula: C₂₂H₃₅BN₂O₄
MW: 402.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16294542
CAS Number: 883738-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR