4'-Methoxybiphenyl-4-ylboronic acid pinacol ester 95%

Synonyms: 2-[4-(4-Methoxyphenyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H51907-03 H51907-06
AAH51907-03EA 63.1 USD
AAH51907-03 AAH51907-06
4'-Methoxybiphenyl-4-ylboronic acid pinacol ester 95%
4'-Methoxybiphenyl-4-ylboronic acid pinacol ester
Formula: C₁₉H₂₃BO₃
MW: 310.20
Storage Temperature: Ambient
MDL Number: MFCD16294530
CAS Number: 446311-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR