Potassium-4-carboxyphenyltrifluoroborate 95%

Warning

Synonyms: Potassium (4-carboxyphenyl)trifluoroborate

H51888-03
AAH51888-03EA 45.04 USD
AAH51888-03
Potassium-4-carboxyphenyltrifluoroborate 95%
Potassium 4-carboxyphenyltrifluoroborate
Formula: C₇H₅BF₃KO₂
MW: 228.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115737
CAS Number: 850623-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR