Potassium-4-(ethoxycarbonyl)phenyltrifluoroborate 95%
|
Warning
|
H51882-03
AAH51882-03EA
45.39
USD
AAH51882-03
Potassium-4-(ethoxycarbonyl)phenyltrifluoroborate 95%
Potassium 4-(ethoxycarbonyl)phenyltrifluoroborate
Formula:
C₉H₉BF₃KO₂ MW: 256.07 Storage Temperature: Ambient |
MDL Number:
MFCD09800743 CAS Number: 1439402-07-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...