2-Hydroxy-3-methylpyrazine 96%

Warning

Synonyms: 3-Methylpyrazin-2-ol3-Methylpyrazin-2(1H)-one

H51880-MD
AAH51880-MDEA 90.53 USD
AAH51880-MD
2-Hydroxy-3-methylpyrazine 96%
2-Hydroxy-3-methylpyrazine
Formula: C₅H₆N₂O
MW: 110.11 g/mol
Melting Pt: 152…153 °C
Storage Temperature: Ambient
MDL Number: MFCD01646120
EINECS: 243-362-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR