2-Chloro-4-fluoro-3-methylbenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H51879-03
H51879-MD
AAH51879-MDEA
113.06
USD
AAH51879-MD
AAH51879-03
2-Chloro-4-fluoro-3-methylbenzoic acid 97%
2-Chloro-4-fluoro-3-methylbenzoic acid
Formula:
C₈H₆ClFO₂ MW: 188.59 g/mol Melting Pt: 186…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD11846444 CAS Number: 173315-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...