6-Fluoro-2,3-dimethoxybenzaldehyde 97%

Warning

H51877-MD
AAH51877-MDEA 100.86 USD
AAH51877-MD
6-Fluoro-2,3-dimethoxybenzaldehyde 97%
6-Fluoro-2,3-dimethoxybenzaldehyde
Formula: C₉H₉FO₃
Melting Pt: 45…47 °C
Storage Temperature: Ambient
MDL Number: MFCD11110608
CAS Number: 457628-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR