2-Iodo-1-tritylimidazole 97%

Supplier: Alfa Aesar
Warning

H51871-MD H51871-03
AAH51871-MDEA 48.91 USD
AAH51871-MD AAH51871-03
2-Iodo-1-tritylimidazole 97%
2-Iodo-1-tritylimidazole
Formula: C₂₂H₁₇IN₂
MW: 436,29
Melting Pt: 170…171 °C
Storage Temperature: Ambient
MDL Number: MFCD02179529
CAS Number: 67478-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR