2-(2-Pyridinyl)-4-thiazolecarboxylic acid 97%

Warning

H51851-03 H51851-06
AAH51851-06EA 293.19 USD
AAH51851-06 AAH51851-03
2-(2-Pyridinyl)-4-thiazolecarboxylic acid 97%
2-(2-Pyridinyl)-4-thiazolecarboxylic acid
Formula: C₉H₆N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD07375352
CAS Number: 115311-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR