3,4-Dimethylthiobenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H51793-03 H51793-MD
AAH51793-03EA 227.99 USD
AAH51793-03 AAH51793-MD
3,4-Dimethylthiobenzamide 97%
3,4-Dimethylthiobenzamide
Formula: C₉H₁₁NS
Storage Temperature: Ambient
MDL Number: MFCD06738326
CAS Number: 58952-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR