4-Aminobenzamidoxime 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-N'-hydroxybenzimidamide

H51788-06 H51788-03
AAH51788-03EA 64.09 USD
AAH51788-03 AAH51788-06
4-Aminobenzamidoxime 97%
4-Aminobenzamidoxime
Formula: C₇H₉N₃O
MW: 151.17 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD08061151
CAS Number: 277319-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR