4-Methyl-2-(3-pyridinyl)-5-thiazolecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 4-Methyl-2-(pyridin-3-yl)thiazole-5-carboxylic acid

H51723-03 H51723-06
AAH51723-03EA 55.9 USD
AAH51723-03 AAH51723-06
4-Methyl-2-(3-pyridinyl)-5-thiazolecarboxylic acid 97%
4-Methyl-2-(3-pyridinyl)-5-thiazolecarboxylic acid
Formula: C₁₀H₈N₂O₂S
MW: 220.25 g/mol
Melting Pt: 244…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00171753
CAS Number: 39091-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR