3-Bromo-5-(trifluoromethyl)phenylboronic acid pinacol ester 97%

Synonyms: 3-Bromo-5-(trifluoromethyl)benzeneboronic acid pinacol ester

H51713-MD H51713-03
AAH51713-03EA 224 USD
AAH51713-03 AAH51713-MD
3-Bromo-5-(trifluoromethyl)phenylboronic acid pinacol ester 97%
3-Bromo-5-(trifluoromethyl)phenylboronic acid pinacol ester
Formula: C₁₃H₁₅BBrF₃O₂
Melting Pt: 50…52 °C
Storage Temperature: Ambient
MDL Number: MFCD12405333
CAS Number: 479411-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR