Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methyl-2-(2-pyridinyl)-5-thiazolecarboxylic acid 97%

Supplier: Alfa Aesar
Warning

H51711-03 H51711-06
AAH51711-03EA 53.87 USD InStock
AAH51711-03 AAH51711-06
4-Methyl-2-(2-pyridinyl)-5-thiazolecarboxylic acid 97%
4-Methyl-2-(2-pyridinyl)-5-thiazolecarboxylic acid


Formula: C₁₀H₈N₂O₂S
MW: 220,25
Melting Pt: 224…228 °C
Storage Temperature: Ambient
MDL Number: MFCD01316364
CAS Number: 34418-48-9
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty