2-Amino-6-iodopurine 97%
H51672-03
H51672-06
AAH51672-03EA
26.5
USD
AAH51672-03
AAH51672-06
2-Amino-6-iodopurine 97%
2-Amino-6-iodopurine
Formula:
C₅H₄IN₅ MW: 261.02 Storage Temperature: Ambient |
MDL Number:
MFCD08706375 CAS Number: 19690-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...