2-(Methylsulfonyl)-5-(tributylstannyl)pyrimidine 96%

Supplier: Thermo Scientific Chemicals

Danger

H51574-MD H51574-03
AAH51574-MDEA 105.56 USD
AAH51574-MD AAH51574-03
2-(Methylsulfonyl)-5-(tributylstannyl)pyrimidine 96%
2-(Methylsulphonyl)-5-(tributylstannyl)pyrimidine
Formula: C₁₇H₃₂N₂O₂SSn
MW: 447.22
Storage Temperature: Ambient
MDL Number: MFCD07787379
CAS Number: 122476-85-1
UN: 2788
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR