1-Methyl-2-(tributylstannyl)indole 96%

Danger

H51515-03
AAH51515-03EA 148.79 USD
AAH51515-03
1-Methyl-2-(tributylstannyl)indole 96%
1-Methyl-2-(tributylstannyl)indole
Formula: C₂₁H₃₅NSn
MW: 420.21
Storage Temperature: Ambient
MDL Number: MFCD09265483
CAS Number: 157427-46-8
UN: 2788
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR