1-Methyl-2-(tributylstannyl)pyrrole 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H51438-MD
H51438-03
AAH51438-03EA
69.69
USD
AAH51438-03
AAH51438-MD
1-Methyl-2-(tributylstannyl)pyrrole 98%
1-Methyl-2-(tributylstannyl)pyrrole
Formula:
C₁₇H₃₃NSn MW: 370.15 Density: 1.122 Flash Pt: 98 °C(208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01319022 CAS Number: 118486-97-8 UN: 2788 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...