1-Methyl-5-(tributylstannyl)imidazole 90+%

Supplier: Thermo Scientific Chemicals

Danger

H51405-03 H51405-MD
AAH51405-MDEA 48.74 USD
AAH51405-MD AAH51405-03
1-Methyl-5-(tributylstannyl)imidazole 90+%
1-Methyl-5-(tributylstannyl)imidazole
Formula: C₁₆H₃₂N₂Sn
MW: 371.15
Storage Temperature: Ambient
MDL Number: MFCD01319031
CAS Number: 147716-03-8
UN: 2788
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR