3-(Tributylstannyl)pyridine 97%

Supplier: Alfa Aesar
Danger

Synonyms: 3-(Tri-n-butylstannyl)pyridine

H51268-03
AAH51268-03EA 35.52 USD
AAH51268-03
3-(Tributylstannyl)pyridine 97%
3-(Tributylstannyl)pyridine
Formula: C₁₇H₃₁NSn
MW: 368,15
Density: 1,153
Storage Temperature: Ambient
MDL Number: MFCD00052054
CAS Number: 59020-10-9
EINECS: 000-000-0
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR