3-(3-Chlorophenoxy)piperidine hydrochloride

Warning

H51180-03 H51180-MD
AAH51180-MDEA 95.93 USD
AAH51180-MD AAH51180-03
3-(3-Chlorophenoxy)piperidine hydrochloride
3-(3-Chlorophenoxy)piperidine hydrochloride
Formula: C₁₁H₁₅Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD11870724
CAS Number: 1185301-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR