1-Heptylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)

Danger

H51173-AP
AAH51173-APEA 273.61 USD
AAH51173-AP
1-Heptylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)
1-Heptylmagnesium chloride
Formula: C₇H₁₅ClMg
MW: 158.95
Storage Temperature: Ambient
MDL Number: MFCD11553268
CAS Number: 42092-01-3
UN: 2924
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR