1-Hexylmagnesium chloride 1 M in MeTHF (2-Methyltetrahydrofuran)

Danger

Synonyms: Hexylmagnesium chloride

H51167-AE H51167-AP
AAH51167-AEEA 159.25 USD
AAH51167-AE AAH51167-AP
1-Hexylmagnesium chloride 1 M in MeTHF (2-Methyltetrahydrofuran)
1-Hexylmagnesium chloride
Formula: C₆H₁₃ClMg
MW: 144.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798087
CAS Number: 44767-62-6
UN: 2924
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR