Potassium-4-(phenylaminocarbonyl)phenyltrifluoroborate 97%

Warning

H51150-03
AAH51150-03EA 33.74 USD
AAH51150-03
Potassium-4-(phenylaminocarbonyl)phenyltrifluoroborate 97%
Potassium 4-(phenylaminocarbonyl)phenyltrifluoroborate
Formula: C₁₃H₁₀BF₃KNO
MW: 303.13
Melting Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD11977719
CAS Number: 912350-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR