5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester

Supplier: Thermo Scientific Chemicals

Synonyms: 1-{[5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl}piperidine

H51117-03 H51117-06
AAH51117-03EA 87.14 USD
AAH51117-03 AAH51117-06
5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester
5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester
Formula: C₁₆H₂₆BNO₂S
MW: 307.26
Storage Temperature: Ambient
MDL Number: MFCD11113036
CAS Number: 1218790-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR