3-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester
|
Warning
|
Synonyms:
Morpholino(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone
, 3-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester
H51113-03
AAH51113-03EA
37.74
USD
AAH51113-03
3-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester
3-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester
Formula:
C₁₇H₂₄BNO₄ MW: 317.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09266172 CAS Number: 1036991-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...