Dimethyl terephthalate-[ring-D4] (98% D)

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: Dimethyl terephthalate-2,3,5,6-d41,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester

H51104-03
AAH51104-03EA 246 USD
AAH51104-03
Dimethyl terephthalate-[ring-D4] (98% D)
Dimethyl terephthalate-[ring-D4]
Formula: C₁₀H₆D₄O₄
Storage Temperature: Ambient
CAS Number: 74079-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR