4-(Methoxymethyl)piperidine hydrochloride
Supplier: Thermo Scientific Chemicals
|
Warning
|
H51020-MD
H51020-03
AAH51020-03EA
279.99
USD
AAH51020-03
AAH51020-MD
4-(Methoxymethyl)piperidine hydrochloride
4-(Methoxymethyl)piperidine hydrochloride
Formula:
C₇H₁₆ClNO Storage Temperature: Ambient |
MDL Number:
MFCD08062602 CAS Number: 916317-00-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...