2-(Methylsulfonyl)benzoic acid 98%

Supplier: Thermo Scientific Chemicals

Warning

H51018-03 H51018-06
AAH51018-03EA 60.05 USD
AAH51018-03 AAH51018-06
2-(Methylsulfonyl)benzoic acid 98%
2-(Methylsulphonyl)benzoic acid
Formula: C₈H₈O₄S
Melting Pt: 136…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00216492
CAS Number: 33963-55-2
EINECS: 223-765-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR